Gene Transcripts

Gene Symbol
Prkar1a
Entrez Gene ID
25725
Gene Name
protein kinase, cAMP-dependent, regulatory, type I, alpha
Gene Aliases
RIIA
Chromosome Location
Chr. 10: 97940705 - 97959199 on Build Rnor_6.0
UniGene ID
Rn.10990
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013181.1 NP_037313.1 8 831
XM_008768311.1 XP_008766533.1 8 887
GenBank mRNA M17086.1 - 8 831
U75932.1 - 5 345

View Full Product Details