Gene Transcripts

Gene Symbol
Grm1
Entrez Gene ID
24414
Gene Name
glutamate receptor, metabotropic 1
Gene Aliases
Gprc1a
Chromosome Location
Chr. 1: 4753144 - 5165859 on Build Rnor_6.0
UniGene ID
Rn.87787
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114330.1 NP_001107802.1 6 1891
NM_017011.1 NP_058707.1 6 1891
GenBank mRNA M61099.1 - 6 1891
X57569.1 - 6 1873

View Full Product Details