Gene Transcripts

Gene Symbol
Srd5a1
Entrez Gene ID
24950
Gene Name
steroid-5-alpha-reductase, alpha polypeptide 1 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1)
Gene Aliases
S5AR 1
Chromosome Location
Chr. 1: 36320504 - 36354694 on Build Rnor_6.0
UniGene ID
Rn.4620
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017070.3 NP_058766.2 670
GenBank mRNA BC127454.1 - 671
EF091857.1 - 156
J05035.1 - 523
S81448.1 - 543

View Full Product Details