Gene Transcripts

Gene Symbol
Cep85l
Entrez Gene ID
100038725
Gene Name
centrosomal protein 85-like
Gene Aliases
ENSMUSG00000038594, Gm9766
Chromosome Location
Chr. 10: 53276799 - 53380701 on Build GRCm38
UniGene ID
Mm.441403
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204983.1 NP_001191912.1 3 798
GenBank mRNA AI642885.1 - 2 367
AK039734.1 - 3 800

View Full Product Details