Gene Transcripts

Gene Symbol
Adora1
Entrez Gene ID
29290
Gene Name
adenosine A1 receptor
Gene Aliases
Chromosome Location
Chr. 13: 51042246 - 51076856 on Build Rnor_6.0
UniGene ID
Rn.32078
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017155.2 NP_058851.2 2 755
XM_006249859.2 XP_006249921.1 3 1046
XM_006249860.2 XP_006249922.1 3 821
XM_006249861.2 XP_006249923.1 3 985
XM_008769552.1 XP_008767774.1 3 1208
XM_008769553.1 XP_008767775.1 3 1147
GenBank mRNA AB001089.1 - 2 692
AF042079.1 - 2 723
M69045.1 - 2 755
Y12519.1 - 2 1107

View Full Product Details