Gene Transcripts

Gene Symbol
Gli2
Entrez Gene ID
14633
Gene Name
GLI-Kruppel family member GLI2
Gene Aliases
AW546128
Chromosome Location
Chr. 1: 118834061 - 119053619 on Build GRCm38
UniGene ID
Mm.273292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081125.1 NP_001074594.1 5, 6 985
GenBank mRNA AM261669.1 - 2, 3 229
AM262742.1 - 4, 5 502
AM262743.1 - 4, 5 599
AM262744.1 - 6, 7 816
BC082604.1 - 5, 6 926 30672479
BC085190.1 - 5, 6 901 30694633
X99104.1 - 5, 6 745

View Full Product Details