Gene Transcripts

Gene Symbol
Pnck
Entrez Gene ID
29660
Gene Name
pregnancy up-regulated nonubiquitous CaM kinase
Gene Aliases
Camk1b
Chromosome Location
Chr. X: 157150078 - 157154558 on Build Rnor_6.0
UniGene ID
Rn.11178
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017275.1 NP_058971.1 5 505
XM_006229572.2 XP_006229634.1 5 337
XM_006229573.2 XP_006229635.1 5 361
XM_006229574.2 XP_006229636.1 5 311
XM_006229575.2 XP_006229637.1 5 315
XM_008773628.1 XP_008771850.1 5 430
XM_008773629.1 XP_008771851.1 4 308
GenBank mRNA AB004267.1 - 5 505
D86556.1 - 5 356

View Full Product Details