Gene Transcripts

Gene Symbol
Cfap44
Entrez Gene ID
212517
Gene Name
cilia and flagella associated protein 44
Gene Aliases
6330444M21Rik, D16Ertd642e, Gm1750, Wdr52
Chromosome Location
Chr. 16: 44394742 - 44482694 on Build GRCm38
UniGene ID
Mm.327888
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033247.1 NP_001028419.1 18 2370
GenBank mRNA AK078103.1 - 14 2569

View Full Product Details