Gene Transcripts
- Gene Symbol
- Ctss
- Entrez Gene ID
- 50654
- Gene Name
- cathepsin S
- Gene Aliases
- Chromosome Location
- Chr. 2: 197655780 - 197679768 on Build Rnor_6.0
- UniGene ID
- Rn.11347
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017320.1 |
NP_059016.1
|
1, 2 |
142 |
|
GenBank mRNA |
L03201.1 |
- |
1, 2 |
142 |
|
View Full Product Details