Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033269.3 | NP_001028441.1 | 7 | 984 | |
NM_001039683.2 | NP_001034772.1 | 7 | 946 | ||
NM_001286178.1 | NP_001273107.1 | 7 | 913 | ||
NM_001286179.1 | NP_001273108.1 | 7 | 916 | ||
NM_001286180.1 | NP_001273109.1 | 7 | 875 | ||
GenBank mRNA | DQ279473.1 | - | 7 | 979 |