Gene Transcripts

Gene Symbol
Uspl1
Entrez Gene ID
231915
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
E430026A01Rik
Chromosome Location
Chr. 5: 149184560 - 149215434 on Build GRCm38
UniGene ID
Mm.29402
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013378.2 NP_001013396.2 10 3016
NM_001115149.1 NP_001108621.1 9 3516
NM_001115150.1 NP_001108622.1 8 3347
NM_001115151.1 NP_001108623.1 9 2847
NM_001115153.1 NP_001108625.1 10 3693
NM_001286824.1 NP_001273753.1 9 3474
NM_001286825.1 NP_001273754.1 9 3458
GenBank mRNA AK088787.1 - 8 3213
AK145409.1 - 10 3591
AK146820.1 - 9 3341
AK161654.1 - 9 2695
AK168702.1 - 9 3341
AK168981.1 - 9 3369
BC028849.1 - 10 2910
BC032291.1 - 7 4986
BC086664.1 - 10 2859

View Full Product Details