Gene Transcripts

Gene Symbol
Mphosph9
Entrez Gene ID
269702
Gene Name
M-phase phosphoprotein 9
Gene Aliases
4930548D04Rik, 9630025B04Rik, AW413446, AW547060, B930097C17Rik, C87456, MPP-9, MPP9
Chromosome Location
Chr. 5: 124250959 - 124330759 on Build GRCm38
UniGene ID
Mm.60224
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081323.2 NP_001074792.1 22 3981
NM_001277867.1 NP_001264796.1 23 4071
GenBank mRNA AK016065.1 - 4 882
BC076624.1 - 22 3844
BC138423.1 - 19 3383 8861443
BC145681.1 - 19 3383 40130984

View Full Product Details