Gene Transcripts

Gene Symbol
Tm4sf19
Entrez Gene ID
277203
Gene Name
transmembrane 4 L six family member 19
Gene Aliases
EG277203
Chromosome Location
Chr. 16: 32400012 - 32408579 on Build GRCm38
UniGene ID
Mm.484240
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160402.1 NP_001153874.1 4, 5 864
GenBank mRNA BC147727.1 - 4, 5 864
BC147729.1 - 4, 5 864 9056678

View Full Product Details