Gene Transcripts

Gene Symbol
A630076J17Rik
Entrez Gene ID
319929
Gene Name
RIKEN cDNA A630076J17 gene
Gene Aliases
Chromosome Location
Chr. 3: 107230614 - 107234031 on Build GRCm38
UniGene ID
Mm.30453
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256174.1 NP_001243103.1 3 416
GenBank mRNA AA119196.1 - 3 393
AK080367.1 - 417

View Full Product Details