Gene Transcripts

Gene Symbol
9330182O14Rik
Entrez Gene ID
328531
Gene Name
RIKEN cDNA 9330182O14 gene
Gene Aliases
Chromosome Location
Chr. 15: 40134365 - 40150010 on Build GRCm38
UniGene ID
Mm.60703
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256056.1 NP_001242985.1 1 72
GenBank mRNA AK034361.1 - 1 74

View Full Product Details