Gene Transcripts

Gene Symbol
Gm5447
Entrez Gene ID
432743
Gene Name
predicted gene 5447
Gene Aliases
EG432743
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.425797
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK077314.1 - 1 1495
BC147407.1 - 1 1452 9056483
BC147408.1 - 1 1455 9056476

View Full Product Details