Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278938.1 | NP_001265867.1 | 3 | 217 | |
NM_004215.4 | NP_004206.1 | 3 | 504 | ||
NM_198120.2 | NP_936056.1 | 3 | 337 | ||
XM_017013960.1 | XP_016869449.1 | 3 | 700 | ||
GenBank mRNA | AB007619.1 | - | 3 | 385 | |
AF006265.1 | - | 3 | 335 | ||
AK290651.1 | - | 3 | 326 | ||
AK308950.1 | - | 3 | 395 | ||
AL515533.3 | - | 3 | 395 | ||
AY515724.1 | - | 3 | 295 | ||
AY653072.1 | - | 2 | 102 | ||
BC005249.1 | - | 3 | 415 | 3930583 | |
BC017729.1 | - | 3 | 288 | 3453385 | |
BC022506.1 | - | 3 | 402 | 4815654 | |
BG616704.1 | - | 3 | 218 | ||
BG707115.1 | - | 3 | 338 | ||
BI194881.1 | - | 3 | 504 | ||
CD108096.1 | - | 3 | 448 | ||
CR456984.1 | - | 2 | 102 |