Gene Transcripts

Gene Symbol
Atxn1l
Entrez Gene ID
52335
Gene Name
ataxin 1-like
Gene Aliases
AW558552, Boat, C330011L24Rik, D6Mgi37, D8Ertd587e, ENSMUSG00000069895
Chromosome Location
Chr. 8: 109726451 - 109737796 on Build GRCm38
UniGene ID
Mm.276770
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080930.1 NP_001074399.1 4 6584
GenBank mRNA AK082766.1 - 1 881

View Full Product Details