Gene Transcripts

Gene Symbol
Amelx
Entrez Gene ID
29160
Gene Name
amelogenin, X-linked
Gene Aliases
Amel
Chromosome Location
Chr. X: 26439197 - 26450495 on Build Rnor_6.0
UniGene ID
Rn.11368
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271074.1 NP_001258003.1 4, 5 214
NM_001271077.1 NP_001258006.1 4, 5 214
NM_019154.1 NP_062027.1 4, 5 205
GenBank mRNA U01245.2 - 3, 4 173
U67130.1 - 4, 5 205

View Full Product Details