Gene Transcripts

Gene Symbol
Defa26
Entrez Gene ID
626708
Gene Name
defensin, alpha, 26
Gene Aliases
Defcr26
Chromosome Location
Chr. 8: 21618168 - 21618972 on Build GRCm38
UniGene ID
Mm.305275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079933.2 NP_001073402.1 2 321
GenBank mRNA CJ043218.1 - 2 322

View Full Product Details