Gene Transcripts
- Gene Symbol
- Ifrd1
- Entrez Gene ID
- 29596
- Gene Name
- interferon-related developmental regulator 1
- Gene Aliases
- Pc4
- Chromosome Location
- Chr. 6: 60132018 - 60151354 on Build Rnor_6.0
- UniGene ID
- Rn.3723
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019242.1 |
NP_062115.1
|
2, 3 |
342 |
|
GenBank mRNA |
J04511.1 |
- |
2, 3 |
342 |
|
View Full Product Details