Gene Transcripts

Gene Symbol
Cst12
Entrez Gene ID
69362
Gene Name
cystatin 12
Gene Aliases
1700006F03Rik, CRES3
Chromosome Location
Chr. 2: 148789361 - 148793437 on Build GRCm38
UniGene ID
Mm.280779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027054.1 NP_081330.1 1 79
GenBank mRNA AF440737.2 - 1 61
AK005670.1 - 1 70
BC119432.2 - 1 79 40041153

View Full Product Details