Gene Transcripts

Gene Symbol
Nkpd1
Entrez Gene ID
69547
Gene Name
NTPase, KAP family P-loop domain containing 1
Gene Aliases
2310015G09Rik
Chromosome Location
Chr. 7: 19516236 - 19525057 on Build GRCm38
UniGene ID
Mm.77325
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027116.1 NP_081392.1 4 2482
GenBank mRNA AK009357.1 - 1 1102
BC118925.2 - 4 2482 40088590

View Full Product Details