Gene Transcripts
- Gene Symbol
- Nkpd1
- Entrez Gene ID
- 69547
- Gene Name
- NTPase, KAP family P-loop domain containing 1
- Gene Aliases
- 2310015G09Rik
- Chromosome Location
- Chr. 7: 19516236 - 19525057 on Build GRCm38
- UniGene ID
- Mm.77325
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027116.1 |
NP_081392.1
|
2, 3 |
547 |
|
GenBank mRNA |
BC118925.2 |
- |
2, 3 |
547 |
|
View Full Product Details