Gene Transcripts

Gene Symbol
Cdk13
Entrez Gene ID
69562
Gene Name
cyclin-dependent kinase 13
Gene Aliases
2310015O17Rik, Cdc2l5
Chromosome Location
Chr. 13: 17715962 - 17805332 on Build GRCm38
UniGene ID
Mm.193924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081058.2 NP_001074527.1 14 6558
NM_027118.1 NP_081394.1 14 6378
GenBank mRNA AK078640.1 - 1 1399
AK144792.1 - 1 393
AK148986.1 - 1 2179

View Full Product Details