Gene Transcripts

Gene Symbol
Utp20
Entrez Gene ID
70683
Gene Name
UTP20, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
3830408P06Rik, AA617408, Drim, mDRIM, mDRIN
Chromosome Location
Chr. 10: 88746607 - 88826814 on Build GRCm38
UniGene ID
Mm.29631
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175158.3 NP_780367.2 39 5123
GenBank mRNA BC048955.1 - 9 988

View Full Product Details