Gene Transcripts

Gene Symbol
4931431F19Rik
Entrez Gene ID
70980
Gene Name
RIKEN cDNA 4931431F19 gene
Gene Aliases
Chromosome Location
Chr. 7: 104127913 - 104129823 on Build GRCm38
UniGene ID
Mm.158518
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027634.1 NP_081910.1 1 96
GenBank mRNA AK016497.1 - 1 97

View Full Product Details