Gene Transcripts

Gene Symbol
Rnf19b
Entrez Gene ID
75234
Gene Name
ring finger protein 19B
Gene Aliases
4930534K13Rik, 4930555L03Rik, Ibrdc3
Chromosome Location
Chr. 4: 129058174 - 129085886 on Build GRCm38
UniGene ID
Mm.259672
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029219.1 NP_083495.1 3 861
GenBank mRNA AK089131.1 - 2 99
BC117807.1 - 3 419
BC139808.1 - 3 422

View Full Product Details