Gene Transcripts

Gene Symbol
1700006E09Rik
Entrez Gene ID
75437
Gene Name
RIKEN cDNA 1700006E09 gene
Gene Aliases
Chromosome Location
Chr. 11: 101987056 - 101992264 on Build GRCm38
UniGene ID
Mm.108632
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029287.1 NP_083563.1 6 658
GenBank mRNA AK005669.1 - 6 662
BB015789.1 - 3 190
BC100517.1 - 6 633

View Full Product Details