Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029393.1 | NP_083669.1 | 1 | 477 | |
GenBank mRNA | AK010165.1 | - | 1 | 502 | |
AK053678.1 | - | 1 | 223 | ||
BC129847.1 | - | 1 | 477 | 40107361 | |
BC129848.1 | - | 1 | 477 | 40107363 | |
GU046702.1 | - | 1 | 446 |