Gene Transcripts

Gene Symbol
Vwa5b1
Entrez Gene ID
75718
Gene Name
von Willebrand factor A domain containing 5B1
Gene Aliases
4931403E03Rik
Chromosome Location
Chr. 4: 138564229 - 138635937 on Build GRCm38
UniGene ID
Mm.251530
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029401.1 NP_083677.1 5 742
GenBank mRNA AK016425.1 - 5 763
BC057376.1 - 4 614
BC132477.1 - 5 742 40130754
BC137851.1 - 5 742 8860870
BC144974.1 - 6 797

View Full Product Details