Gene Transcripts

Gene Symbol
Zfp773
Entrez Gene ID
76373
Gene Name
zinc finger protein 773
Gene Aliases
2810409K11Rik
Chromosome Location
Chr. 7: 7127073 - 7136785 on Build GRCm38
UniGene ID
Mm.159732
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029584.1 NP_083860.1 1 133
GenBank mRNA AK013056.1 - 1 101
AK031040.1 - 1 133
AK036639.1 - 1 103
BC117497.1 - 1 111 40106595

View Full Product Details