Gene Transcripts

Gene Symbol
Bmp15
Entrez Gene ID
59302
Gene Name
bone morphogenetic protein 15
Gene Aliases
Gdf-9b
Chromosome Location
Chr. X: 17016831 - 17021894 on Build Rnor_6.0
UniGene ID
Rn.42867
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021670.1 NP_067702.1 1 451
GenBank mRNA AJ132407.1 - 1 451

View Full Product Details