Gene Transcripts
- Gene Symbol
- Htr1f
- Entrez Gene ID
- 60448
- Gene Name
- 5-hydroxytryptamine (serotonin) receptor 1F, G protein-coupled
- Gene Aliases
- Htr1eb
- Chromosome Location
- Chr. 11: 1982113 - 1983513 on Build Rnor_6.0
- UniGene ID
- Rn.44301
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021857.3 |
NP_068629.2
|
1 |
542 |
|
View Full Product Details