Gene Transcripts

Gene Symbol
Sh3bp4
Entrez Gene ID
64634
Gene Name
SH3-domain binding protein 4
Gene Aliases
Chromosome Location
Chr. 9: 96210750 - 96288903 on Build Rnor_6.0
UniGene ID
Rn.35396
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022693.4 NP_073184.3 4 801
XM_008767298.1 XP_008765520.1 2 1447
GenBank mRNA AJ278266.2 - 2 476

View Full Product Details