Gene Transcripts

Gene Symbol
Bche
Entrez Gene ID
65036
Gene Name
butyrylcholinesterase
Gene Aliases
Chromosome Location
Chr. 2: 171100137 - 171196186 on Build Rnor_6.0
UniGene ID
Rn.48791
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022942.1 NP_075231.1 2 935
XM_008761112.1 XP_008759334.1 1 1012
GenBank mRNA AF244349.1 - 2 935

View Full Product Details