Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024159.1 | NP_077073.1 | 1, 2 | 105 | |
GenBank mRNA | AF045657.1 | - | 1, 2 | 95 | |
BC097314.1 | - | 1, 2 | 140 | 7444786 | |
FQ224185.1 | - | 415 | |||
U95177.1 | - | 1, 2 | 105 | ||
U95178.1 | - | 1, 2 | 105 |