Gene Transcripts

Gene Symbol
Cdk5rap3
Entrez Gene ID
80278
Gene Name
CDK5 regulatory subunit associated protein 3
Gene Aliases
C53
Chromosome Location
Chr. 10: 84780646 - 84789787 on Build Rnor_6.0
UniGene ID
Rn.3727
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024488.1 NP_077814.1 5, 6 413
GenBank mRNA AF177476.1 - 5, 6 413
BC081793.1 - 4, 5 335 7190258

View Full Product Details