Gene Transcripts
- Gene Symbol
- Mvk
- Entrez Gene ID
- 81727
- Gene Name
- mevalonate kinase
- Gene Aliases
- Lrbp
- Chromosome Location
- Chr. 12: 47904266 - 47920457 on Build Rnor_6.0
- UniGene ID
- Rn.6995
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031063.1 |
NP_112325.1
|
6 |
626 |
|
GenBank mRNA |
M29472.1 |
- |
6 |
626 |
|
View Full Product Details