Gene Transcripts

Gene Symbol
Stxbp2
Entrez Gene ID
81804
Gene Name
syntaxin binding protein 2
Gene Aliases
Munc18b
Chromosome Location
Chr. 12: 2180103 - 2191863 on Build Rnor_6.0
UniGene ID
Rn.10121
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031126.1 NP_112388.1 2, 3 101
XM_006248783.2 XP_006248845.1 2, 3 148
GenBank mRNA BC097310.1 - 2, 3 124
U20283.1 - 2, 3 101

View Full Product Details