Gene Transcripts
- Gene Symbol
- Oprl1
- Entrez Gene ID
- 29256
- Gene Name
- opiate receptor-like 1
- Gene Aliases
- KOR-3, KOR3, LC132, MOR-C, OFQR, ORL1, Oprl, XOR1
- Chromosome Location
- Chr. 3: 177225366 - 177231663 on Build Rnor_6.0
- UniGene ID
- Rn.9762
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031569.3 |
NP_113757.1
|
2, 3 |
473 |
|
XM_006235736.2 |
XP_006235798.1
|
3, 4 |
563 |
|
XM_006235738.2 |
XP_006235800.1
|
2, 3 |
321 |
|
XM_006235739.2 |
XP_006235801.1
|
4, 5 |
597 |
|
XM_008762474.1 |
XP_008760696.1
|
1, 2 |
83 |
|
GenBank mRNA |
AF115267.1 |
- |
2, 3 |
294 |
|
AY152731.1 |
- |
1, 2 |
232 |
|
D16438.1 |
- |
2, 3 |
284 |
|
L28144.1 |
- |
2, 3 |
404 |
|
L29419.1 |
- |
1, 2 |
367 |
|
L33916.1 |
- |
2, 3 |
338 |
|
U01913.1 |
- |
2, 3 |
413 |
|
U05239.1 |
- |
2, 3 |
415 |
|
U07871.1 |
- |
1, 2 |
263 |
|
View Full Product Details