Gene Transcripts

Gene Symbol
Ctsk
Entrez Gene ID
29175
Gene Name
cathepsin K
Gene Aliases
Chromosome Location
Chr. 2: 196655469 - 196666447 on Build Rnor_6.0
UniGene ID
Rn.5598
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031560.2 NP_113748.1 4 454
GenBank mRNA AF010306.1 - 4 423
BC078793.1 - 4 454 7134253
FQ222438.1 - 4 398
FQ228691.1 - 4 471

View Full Product Details