Gene Transcripts

Gene Symbol
Ighmbp2
Entrez Gene ID
29532
Gene Name
immunoglobulin mu binding protein 2
Gene Aliases
AEP
Chromosome Location
Chr. 1: 218509274 - 218531922 on Build Rnor_6.0
UniGene ID
Rn.23984
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031586.1 NP_113774.1 5, 6 708
GenBank mRNA AF199411.1 - 5, 6 708
BC099790.1 - 5, 6 904 7934952

View Full Product Details