Gene Transcripts

Gene Symbol
Slc7a7
Entrez Gene ID
83509
Gene Name
solute carrier family 7 (amino acid transporter light chain, y+L system), member 7
Gene Aliases
y+LAT1
Chromosome Location
Chr. 15: 33013353 - 33059739 on Build Rnor_6.0
UniGene ID
Rn.163294
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031341.1 NP_112631.1 2 389
XM_006252020.2 XP_006252082.1 2 325
XM_006252021.2 XP_006252083.1 5 794
XM_006252023.2 XP_006252085.1 2 330
XM_006252024.2 XP_006252086.1 2 1019
XM_008770705.1 XP_008768927.1 4 737
GenBank mRNA AB020520.1 - 2 389
AF200684.1 - 1 197
BC091142.1 - 2 304 7129657

View Full Product Details