Gene Transcripts

Gene Symbol
Gria1
Entrez Gene ID
50592
Gene Name
glutamate receptor, ionotropic, AMPA 1
Gene Aliases
GluA1, gluR-A
Chromosome Location
Chr. 10: 42441723 - 42760200 on Build Rnor_6.0
UniGene ID
Rn.29971
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031608.1 NP_113796.1 2 316
GenBank mRNA M36418.2 - 2 119
M38060.1 - 2 119
X17184.1 - 2 316

View Full Product Details