Gene Transcripts

Gene Symbol
Sh2b3
Entrez Gene ID
58838
Gene Name
SH2B adaptor protein 3
Gene Aliases
Lnk
Chromosome Location
Chr. 12: 40261990 - 40265757 on Build Rnor_6.0
UniGene ID
Rn.11228
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031621.1 NP_113809.1 1 456
GenBank mRNA U24654.1 - 3 303
U24655.1 - 1 456

View Full Product Details