Gene Transcripts
- Gene Symbol
- Polr1b
- Entrez Gene ID
- 83582
- Gene Name
- polymerase (RNA) I polypeptide B
- Gene Aliases
- RPA2, Rpo1-2
- Chromosome Location
- Chr. 3: 121632043 - 121656506 on Build Rnor_6.0
- UniGene ID
- Rn.88734
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031773.1 |
NP_113961.1
|
3, 4 |
558 |
|
GenBank mRNA |
AF025424.1 |
- |
3, 4 |
558 |
|
View Full Product Details