Gene Transcripts

Gene Symbol
Mdh1
Entrez Gene ID
24551
Gene Name
malate dehydrogenase 1, NAD (soluble)
Gene Aliases
MDL1, Mdhl, Mor2
Chromosome Location
Chr. 14: 106378945 - 106393603 on Build Rnor_6.0
UniGene ID
Rn.13492
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033235.1 NP_150238.1 4 271
GenBank mRNA AF093773.1 - 4 271
BC059124.1 - 4 312 6921518
FQ212702.1 - 4 315
FQ215129.1 - 4 325
FQ215603.1 - 4 310
FQ217383.1 - 4 315
FQ217819.1 - 315
FQ224891.1 - 4 313
FQ233582.1 - 4 315

View Full Product Details