Gene Transcripts

Gene Symbol
Ogfr
Entrez Gene ID
83525
Gene Name
opioid growth factor receptor
Gene Aliases
Chromosome Location
Chr. 3: 176093662 - 176099949 on Build Rnor_6.0
UniGene ID
Rn.15740
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053340.2 NP_445792.2 2 186
GenBank mRNA BC101898.1 - 2 167
EV778659.1 - 2 186

View Full Product Details