Gene Transcripts

Gene Symbol
Hdac3
Entrez Gene ID
84578
Gene Name
histone deacetylase 3
Gene Aliases
Chromosome Location
Chr. 18: 31073057 - 31094347 on Build Rnor_6.0
UniGene ID
Rn.17284
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053448.1 NP_445900.1 290
GenBank mRNA AF321131.1 - 290
BC061988.1 - 293 5599517

View Full Product Details